TMO362150-5130
Nalgene® red caps
polypropylene cap, diam. × thread 16 mm × 13-414 mm, case of 2000 ea
Synonym(e):
bottle cap, bottle closure
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Empfohlene Produkte
Materialien
polypropylene cap
red cap
Verpackung
case of 2000 ea
Hersteller/Markenname
Nalgene 362150-5130
Durchm. × Gewinde
16 mm × 13-414 mm
Verwandte Kategorien
Allgemeine Beschreibung
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)