Merck
TMO362089-0016
Nalgene® narrow-mouth bottle without cap
natural high-density polyethylene, volume 500 mL, case of 125 ea
Synonym(e):
Nalgene bottle, plastic bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
natural high-density polyethylene
Verpackung
case of 125 ea
Hersteller/Markenname
Nalgene 362089-0016
Kapazität
500 mL
Volumen
500 mL
Allgemeine Beschreibung
Safely store and handle test kits and many small volume applications with Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Rechtliche Hinweise
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)