Merck
TMO267566
Nunc® Edge 2.0 96-well Plate
untreated, polystyrene, non-sterile, without lid, clear, case of 50 ea
Synonym(e):
cell culture plate, cell growth plate
About This Item
UNSPSC-Code:
41122102
NACRES:
NB.15
Materialien
clear
polystyrene
Sterilität
non-sterile
Verpackung
case of 50 ea
Hersteller/Markenname
Nunc 267566
Allgemeine Beschreibung
Minimize plate evaporation and the risks associated with edge effect by using the uniquely engineered Thermo Scientific Nunc Edge 2.0 Plate. The surrounding bulit-in reservoirs, when filled with sterile water or media, act as evaporation barriers, yielding consistency across the entire 96-well plate. Enabling the use of all 96 wells in a plate allows researchers to plan experiments more efficiently with enhanced productivity and less waste for cell-based assays that require extended incubation.
Rechtliche Hinweise
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)