MTCR1040O
MTC™ Bio Fraction CollectorTube Rack
holds 80 x 1.5 /2.0 mL tubes, orange, pack of 5 ea
Synonym(e):
multi gel processing box, tube holder, tube storage
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122804
Materialien
orange
Leistungsmerkmale
holds 80 x 1.5 /2.0 mL tubes
Verpackung
pack of 5 ea
Hersteller/Markenname
MTC™ Bio R1040-O
Verwandte Kategorien
Allgemeine Beschreibung
Tube Rack, fraction collector type, 80 x 1.5mL/2mL, red, pack of 5
Leistungsmerkmale und Vorteile
- 80 well (5 rows of 16) for 1.5 to 2.0mL microtubes
- Rainbow pack available — 5 colors
- Two frosted marking areas, on the bottom and side
- Raised numbers beside each well
- Recessed well bottoms for improved tube stability
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)