MTCP521250U
MTC™ Bio ProPette™ LE 12-Channel Adjustable Pipettor
capacity 5-50 μL
Synonym(e):
pipette
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Empfohlene Produkte
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC™ Bio P5212-50U
Kapazität
5-50 μL
Allgemeine Beschreibung
ProPette™ LE 12 -Channel Pipette, 5 to 50µL
Leistungsmerkmale und Vorteile
- Adjustable with Roto-Lock
- Light weight with reduced forces
- Full volume range (1.0µL-300µL)
- Superior accuracy and reproducibility
- Outstanding user comfort
Rechtliche Hinweise
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio