MTCP520810U
MTC™ Bio ProPette™ LE 8-Channel Adjustable Pipettor
capacity 1-10 μL
Synonym(e):
pipette
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Empfohlene Produkte
Leistungsmerkmale
adjustable
manual
no. of channels 8
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC™ Bio P5208-10U
Kapazität
1-10 μL
Allgemeine Beschreibung
Ideal for all 96-well applications, Propette LE™ Multi-Channel Pipettes are lightweight, rugged, and capable of standing up to the demands of everyday use in a busy laboratory environment.
ProPette™ LE 8 -Channel Pipette, 1 to 10µL
Leistungsmerkmale und Vorteile
- Adjustable with Roto-Lock
- Light weight with reduced forces
- Full volume range (1.0µL-300µL)
- Superior accuracy and reproducibility
- Outstanding user comfort
Rechtliche Hinweise
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)