Merck
MTCP3802Q
MTC Bio Pure.AmpqPCR 8 Tube Strips
capacity 0.2 mL, clear tube, clear cap (optical), flat cap
Synonym(e):
PCR sample storage tubes, plastic tube, sample storage tube
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.15
Materialien
clear cap (optical)
clear tube
flat cap
polypropylene
Sterilität
non-sterile
Verpackung
pack of 120 ea
Hersteller/Markenname
MTC Bio P3802-Q
Kapazität
0.2 mL
Allgemeine Beschreibung
0.2ml qPCR 8-Strip (With SEPARATE Optical Strip Caps), Natural/Clear, 120/pk
Pure.Amp 0.1mL qPCR strips strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)