Merck
MTCP3020
MTC Bio Pure.Amp PCR 8 Tube Strips
capacity 0.2 mL, seperate dome caps
Synonym(e):
PCR sample storage tubes, sample storage tube, snap closure tube
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.15
Materialien
(self standing star foot)
clear
polypropylene
Sterilität
non-sterile
Verpackung
pack of 120 ea
Hersteller/Markenname
MTC Bio P3020
Kapazität
0.2 mL
Allgemeine Beschreibung
PCR Tubes Strips of 8 tubes w/ dome caps packaged separately, 120/pk
PureAmp 0.2mL PCR strips can be supplied with domed caps or flat caps and can be attached or supplied separately. Optically clear caps and white tubes (for optimal fluorescence reflectivity) are also available for qPCR applications. PureAmp 0.1mL PCR strips are intended for qPCR applications and come in Clear, White, and Frosted. Strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Leistungsmerkmale und Vorteile
  • RNase, DNase, DNA, and PCR inhibitor free
  • 0.2mL or Low Profile 0.1mL options
  • Ultra-thin, uniform wall thickness
  • White and frosted tubes for enhanced qPCR results
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)