Merck
MTCH6800SK
MTC Bio Halo Lightweight Single Channel Pipettor Starter Kit
4 pipettors and a 6 place carousel stand
Synonym(e):
pipette
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.15
Leistungsmerkmale
adjustable
manual
no. of channels 1
Verpackung
pack of 1 ea
Hersteller/Markenname
MTC Bio H6800-SK
Verwandte Kategorien
Allgemeine Beschreibung
Halo Pipettor Starter Kit, includes H6800-10 (10µL), H6800-20 (20µL), H6800-200 (200µL), H680-1MXL (1250µL), and H6800-CR (6 place carousel stand)
Leistungsmerkmale und Vorteile
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Rechtliche Hinweise
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)