Merck
MTCC2625R
MTC Bio Screw Cap Centrifuge Tube
capacity 25 mL, conical bottom, sterile, pack of 200 ea (8 x foam racks 25 ea)
Synonym(e):
conical tube, plastic tube
About This Item
UNSPSC-Code:
41121703
NACRES:
NB.78
Materialien
blue cap
clear tube
conical bottom
polypropylene
round
screw top cap
Sterilität
sterile
Verpackung
pack of 200 ea (8 x foam racks 25 ea)
Hersteller/Markenname
MTC Bio C2625-R
Parameter
12,000 × g speed (max)
Kapazität
25 mL
Durchm. × L
27 mm × 78 mm
Allgemeine Beschreibung
Centrifuge Tube, 25mL, PP, sterile, with screw cap, 8 foam racks of 25 tubes, 200/pack
These centrifuge tubes are made of premium polypropylene to maximize centrifugal strength and maintain clarity. Tubes and caps may be frozen at temperatures as low as 86˚C. Caps have a flat top surface for easy marking and labeling.
Leistungsmerkmale und Vorteile
  • Sterile, RNase, DNase, pyrogen and cytotoxin free
  • Available racked or bagged
  • Compatible with standard rotors
  • Frosted marking surface
Rechtliche Hinweise
MTC is a trademark of MTC Bio
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)