Merck
HS20632B
Pipette Filler
red natural rubber
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
Materialien
red natural rubber
Verpackung
pack of 1 ea
Hersteller/Markenname
Heathrow Scientific HS20632B
L × B × H
13 cm × 5.4 cm × 5.4 cm
Kapazität
~0.35 mL
Allgemeine Beschreibung
The natural rubber Pipette Filler is great for use in one-handed operation.
Leistungsmerkmale und Vorteile
  • Efficient one-handed operation
  • Control air-evacuation, liquid uptake, and liquid dispensing with three valves each containing chemical-resistant stainless steel balls in valves
  • Pinch release-valve for smooth liquid flow
  • 5.1 in L x 2.1 in W x 2.1 in H (13 cm L x 5.4 cm W x 5.4 cm H)
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)