Merck
HS120817
OneRack® Multi
blue polypropylene (/natural)
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.12
Materialien
blue polypropylene (/natural)
Leistungsmerkmale
to hold 20 x 10-17 mm tubes OR 12 x 25 - 30 mm tubes
Verpackung
pkg of 1 ea
Hersteller/Markenname
Heathrow Scientific 120817
L × B × H
309 mm × 107 mm × 77 mm
Well-Durchm. × Durchm. × Durchm.
10-13 mm × 13-17 m2 × 25-30 mm
Allgemeine Beschreibung
The OneRack® Multi is the most flexible tube rack designed for use with a multitude of tube diameters and heights. Each rack comes with 1 base and 6 inserts. Of the 6 inserts, there are two of each size (10-13mm, 13-17mm, 25-30 mm).
Leistungsmerkmale und Vorteile
  • Interchangeable tube rack inserts hold multiple tube sizes firmly in place when alternating between sample vessels
  • Inserts can be pivoted in either an upper or lower position to securely accommodate the size and height of tube being used
  • Open design allows for better filling visibility of tube graduations when filling
  • Rack wells have a unique teardrop design that enables better retention of tubes over traditional designs
  • Able to rotate the tube rack insert to an angle position to facilitate more comfortable pipetting
Rechtliche Hinweise
OneRack is a registered trademark of Heathrow Scientific LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)