Merck
HS120618
Premium Plus Slide Box replacement inventory cards
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.16
Materialien
cardboard
Verpackung
pack of HS120618
Hersteller/Markenname
Heathrow Scientific 120618
L × B × H
168.3 mm × 0.5 mm × 203.2 mm
Allgemeine Beschreibung
Premium Plus Slide Box replacement inventory cards designed to fit into the premium plus slide boxes.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)