GBL622201
Grace Bio-Labs CoverWell™ perfusion chambers
1 chamber, port diam. 1.5 mm, chamber diam. × thickness 20 mm × 1.7 mm
Synonym(e):
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
About This Item
Empfohlene Produkte
Materialien
clear polycarbonate backing
red silicon wells
Leistungsmerkmale
wells 1
Verpackung
pack of 20 ea
Hersteller/Markenname
Grace Bio-Labs 622201
Kammerdurchm. × Dicke
20 mm × 1.7 mm
Öffnungsdurchm.
1.5 mm
Wells
1
Verwandte Kategorien
Allgemeine Beschreibung
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
GBL622201-20EA | 4061832399423 |