Merck
EPM1282-9926
Platform for Eppendorf® New Brunswick Innova® 44/44R Incubator Shakers
for 2 L Erlenmeyer flasks, dedicated, non-Interchangeable, aluminum (heavy duty)
About This Item
UNSPSC-Code:
41103816
NACRES:
NB.73
Materialien
aluminum (heavy duty)
Leistungsmerkmale
dedicated
for 2 L Erlenmeyer flasks
non-Interchangeable
Hersteller/Markenname
Eppendorf® M1282-9926 (Global)
Kapazität
(12 x 2 L Erlenmeyer flasks)
Verwandte Kategorien
Allgemeine Beschreibung
Platforms to fit the Eppendorf New Brunswick 44 and 44 R incubated shakers.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc. 
New Brunswick is a trademark of Eppendorf AG