Merck
EP5910704003
Adapter for Eppendorf® S-4x400 Rotor
for 16x75 mm-100 mm round-bottom tubes, pkg of 2 ea
About This Item
UNSPSC-Code:
41103911
NACRES:
NB.82
Leistungsmerkmale
for 16x75 mm-100 mm round-bottom tubes
Verpackung
pkg of 2 ea
Hersteller/Markenname
Eppendorf® 5910704003
Methode(n)
centrifugation: suitable
Kompatibilität
for use with 400 mL round bucket
Allgemeine Beschreibung
Adapter for various size tubes and bottles for use with Eppendorf S-4x400 rotor.
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG