Merck
EP5810729009
Eppendorf® Wide-neck bottle
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.82
Materialien
polypropylene bottle
polypropylene cap
round bottle
Sterilität
non-sterile
Leistungsmerkmale
graduations: no
with cap
Verpackung
pack of 2 ea
Hersteller/Markenname
Eppendorf® 5810729009
Kapazität
400 mL
Allgemeine Beschreibung
400 mL wide-neck bottle which are plastic vessels for preparing, transporting, storing and centrifuging solid and liquid samples and reagents. When centrifuging at high rotational speeds or high temperatures, push the supplied sleeve over the wide-neck bottle. The 400 mL wide-neck bottle does not fit under the cap of the rectangular bucket for therotor A-4-81. It is autoclavable (121C, 20 mins).
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)