EP5392070011
Eppendorf(R) Plate Adapter HeatSealer S200
for 96 and 384 well plates, high profile
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.15
Empfohlene Produkte
Leistungsmerkmale
for 96 and 384 well plates
high profile
Hersteller/Markenname
Eppendorf® 5392070011 (Global)
Verwandte Kategorien
Leistungsmerkmale und Vorteile
Hermetic Sealing, of multiwell plates
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Eliminates evaporation in PCR
Seals 96- and 384-well plates
Integrated thermostat prevents overheating
Improved mechanics for even easier Sealing,
PCR Plate Adapter included
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)