Merck
EP0030124820
Eppendorf® PCR tube strips, 0.1 mL
with cap strips, flat, 0.1 mL, PCR clean, capacity 0.1 mL, with Cap Strips, flat (10x12 strips), pkg of 1000 ea
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.11
Leistungsmerkmale
PCR clean
with Cap Strips, flat (10x12 strips)
Verpackung
pkg of 1000 ea
Hersteller/Markenname
Eppendorf® 0030124820
Kapazität
0.1 mL
0.1 mL
Leistungsmerkmale und Vorteile
  • 8 reaction tubes in strip-format – ideal for small sample volumes
  • Easily adaptable for automation
  • Sealable using flat or domed strip-lids
  • Certified free from human DNA, DNase, RNase and PCR inhibitors
Rechtliche Hinweise
Eppendorf is a registered trademark of Eppendorf AG