Merck
EMU073081
Sigma-Aldrich
MISSION® esiRNA
targeting mouse Tspan1
About This Item
UNSPSC-Code:
41105324
NACRES:
NA.51
Beschreibung
Powered by Eupheria Biotech
Qualitätsniveau
Produktlinie
MISSION®
Form
lyophilized powder
esiRNA cDNA-Zielsequenz
GCAGTTTCCTGGAGCTGTTCTGTTCCTTCTGGAGTGCAACAGATCCCGTTCAGAACCTATTGCCAAGAGCCCTGAAGCAGGACACACCATGCAGTGCTTCAAATTCATTAAGGTCATGATGTTCCTCTTCAATCTACTCATCTTTCTCTGTGGTGCAGCCCTGTTGGCTGTGGGAATCTGGGTGTCCGTCGATGGGACATCTTTCCTGAAGGTCTTCGGATCACTATCATCCAGTGCCATGCAGTTTGTCAACGTGGGCTACTTCCTCATCGCCGCTGGTGCTGTGCTCTTCATTCTTGGTTTCCTGGGCTGCTATGGTGCTCACTCTGAGAACAAGTGTGTGCTCATGATGTTCTTTTCCATCCTCCTCATCATCTTCATCGCTGAGATTGCAGGTGCTGTGGTTGCTTTGGTGTACACCACATTGGCTGAACAA
Ensembl | Maus Hinterlegungsnummer
NCBI-Hinterlegungsnummer
Versandbedingung
ambient
Lagertemp.
−20°C
Angaben zum Gen
Allgemeine Beschreibung
MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Rechtliche Hinweise
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Lagerklassenschlüssel
10 - Combustible liquids
Flammpunkt (°F)
Not applicable
Flammpunkt (°C)
Not applicable
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)