CLS766119
Pyrex® pennyhead stopper
stopper joint: 19
Synonym(e):
glass stopper, glass stoppers, stoppers, Corning® 7660
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122500
NACRES:
NB.43
Empfohlene Produkte
Materialien
glass
Verbindung
joint: stopper19
Hersteller/Markenname
Corning 7661-19
Allgemeine Beschreibung
Ground zone length indicates its greatest diameter (mm).
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)