CLS2975243
Corning® cover glasses
rectangular, No. 1, W × L 24 mm × 30 mm
Synonym(e):
coverslips, glass covers, slide covers
About This Item
Empfohlene Produkte
Materialien
borosilicate glass
rectangular
Verpackung
pack of 100 ea
case of 1000 ea
Hersteller/Markenname
Corning 2975-243
B × L
24 mm × 30 mm
Bindungsart
non-treated surface
Verwandte Kategorien
Leistungsmerkmale und Vorteile
- Corning® cover glasses are made from optically clear borosilicate glass.
- Chemically resistant cover glasses are not affected by weathering or surface attachment.
Rechtliche Hinweise
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CLS2975243-1000EA | 4061833882382 |
CLS2975243-100EA | 4061833882399 |