Merck
CLS11000
Corning® HYPERStack® stainless steel manipulator
accommodates both HYPERStack-36 and CellSTACK-10 vessels
Synonym(e):
lab equipment accessory
About This Item
UNSPSC-Code:
41122800
NACRES:
NA.75
Hersteller/Markenname
Corning
Allgemeine Beschreibung
  • Made of stainless steel for durability
  • Accommodates both HYPERStack-36 and CellSTACK-10 vessels
  • Ergonomic design
  • Easy dials for protocol development
  • Handles two units simultaneously or one unit at a time
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
HYPERStack is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)