BMSW10051
Accuris™ M1 Grade Calibration Weight
size 1 g, stainless steel
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.71
Empfohlene Produkte
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
Benchmark Scientific W1005-1
Größe
1 g
Verwandte Kategorien
Allgemeine Beschreibung
- F1 Grade tolerance per OIML (International Organization of Legal Metrology)
- Works with all Accuris Lab balances
- Non-Magnetic Stainless Steel
- Includes a protective plastic case
Rechtliche Hinweise
Accuris is a trademark of Benchmark Scientific, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)