BAH378620102
SP Bel-Art®Black Pen
Black Felt Tip Pen
Alle Fotos(3)
About This Item
UNSPSC-Code:
41104000
NACRES:
NB.22
Empfohlene Produkte
Hersteller/Markenname
Bel-Art H37862-0102
Allgemeine Beschreibung
A cost effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate.
Leistungsmerkmale und Vorteile
- Fast, accurate and economical sample counting and sample count verification
- Marks, counts, and gives an audible and visual alert each time a count is registered
- Lightweight, pen-style design reduces the likelihood of error; combines all aspects of counting into one motion
- Stores up to 35 sample counts
- Verify sample counts with up/down counting
- Displays sum total of all colonies counted
- Backlit display for easy viewing in low light conditions
- Audible alert can be turned off if desired
- Counting function can be locked to prevent error during breaks between counting
- Long life, red and black felt-tip markers (included) work on plastic or glass and are oil and water resistant; ink is removable with ethyl alcohol
- Includes 3V lithium battery and protective storage case
- For a complete Colony Counter System, see catalog no. F37863-3000
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)