Merck
BAF243040000
Vikem® Vinyl Microscope Cover
Extra Large, size 22 × 13 × 22 in.
About This Item
UNSPSC-Code:
41122600
NACRES:
NB.22
Materialien
vinyl
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F24304-0000
Größe
22 × 13 × 22 in.
Allgemeine Beschreibung
Vikem vinyl covers easily slide over valuable microscopes, balance and other instruments when not in use.
  • Protects instruments from dust and spills
  • Transparency permits identification of equipment without removing cover
Rechtliche Hinweise
Vikem is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)