AXYRESSW96HPSI
AxyVoir reservoir
96 bottom, high profile single well, individually wrapped, 25 ea of, sterile
Synonym(e):
storage reservoir
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121522
NACRES:
NB.22
Empfohlene Produkte
Sterilität
sterile
Hersteller/Markenname
Axygen Inc RES-SW96-HP-SI
Corning
Verpackung
25 ea of
Verwandte Kategorien
Allgemeine Beschreibung
- Axygen® reagent reservoir is a multiple-well high-profile reservoir manufactured using chemically resistant polypropylene.
- Ideal for both manual and automated applications
- Designed with standard ANSI/SLAS footprint dimensions for microplates.
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)