AXYP96450RY
Round bottom assay plate
size 96 wells, round bottom yellow polypropylene wells, maximum volume 500 μL, pkg of 10x5plates/cs
Synonym(e):
96 multiwell plate
About This Item
Materialien
round bottom yellow polypropylene wells
yellow plate
Sterilität
non-sterile
Hersteller/Markenname
Axygen Inc P-96-450R-Y
Corning
Verpackung
pkg of 10x5plates/cs
Maximales Volumen
500 μL
Größe
96 wells
Arbeitsvolumen
450 μL
Bindungsart
non-treated surface