AXYHGB101615
Corning® Axygen® 16 well comb for use with 10 cm gel system
thickness (1.5 mm)
Synonym(e):
gel system accessory
Alle Fotos(2)
About This Item
UNSPSC-Code:
41105300
NACRES:
NB.15
Empfohlene Produkte
Hersteller/Markenname
Corning HGB10-16-15
Dicke
(1.5 mm)
Verwandte Kategorien
Rechtliche Hinweise
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
AXYHGB101615 |