Wichtige Dokumente
NA38/25
Millipore
Diaphragm for NOVASEPTIC® Valve
PTFE
About This Item
UNSPSC-Code:
31411800
Materialien
PTFE
PTFE device (polytetrafluoroethylene)
Qualitätsniveau
Leistungsmerkmale
hydrophobic (Surface)
Parameter
-40-170 °C temp. range (Continous use)
-40-250 °C temp. range (Short term use)
Gewicht
0.09 kg (0.20 lb)
Anwendung
The applied valve body and diaphragm may have different design temperature and/or pressure limits. The weakest component in the assembled product determines the maximum design temperature and pressure limits.
Verpackung
The Diaphragm is packaged in a closed box.
Sonstige Hinweise
All Diaphragms are manufactured with materials in compliance with FDA regulations §177.2600 and supplied with statement. The Diaphragm meets USP Biological Tests for Plastics, Class VI.
Rechtliche Hinweise
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany