MFSL06SH3
Durapore® 5.0 µm, Millipak® Final Fill Capsule
Millipak® Final Fill 60
About This Item
Materialien
PVDF membrane
polysulfone housing
silicone O-ring
Sterilität
sterile; irradiated
Produktlinie
Millipak® Final Fill 60
Leistungsmerkmale
hydrophilic
Methode(n)
prefiltration: suitable
Matrix
Durapore®
Porengröße
5.0 μm pore size
Fitting
fitting (6 mm (1/4 in.) Hose Barb Inlet and Outlet)