Merck
Z748110
Clamping rings for glove box ports
port O.D. 8 in.
Synonym(e):
humidity chambers clips
About This Item
UNSPSC-Code:
42132201
NACRES:
NB.35
Verpackung
pkg of 1 ea (1 ring per bag)
Hersteller/Markenname
Bel-Art H50025-0320
Öffnung AD
8 in.
Allgemeine Beschreibung
Compatible with Scienceware Economy (T50024-series) and sidENTRY Glove Boxes.
  • Ring is adjustable
  • Secures glove firmly to glove box
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)