Z743199
Julabo® bath test tube racks
to fit 90 test tubes, diam. 12 mm (13 mm), stainless steel
Synonym(e):
tube holder
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.77
Empfohlene Produkte
Materialien
stainless steel
Verpackung
pack of 1 ea
Hersteller/Markenname
Julabo 8970345
Parameter
100 °C max. temp.
Durchmesser
12 mm (13 mm)
Verwandte Kategorien
Allgemeine Beschreibung
Stainless steel rack suitable for temperatures up to 100°C. If solvent based samples or acidic samples are tempered above 60°C, it is recommended using test glass inserts made of stainless steel.
Rechtliche Hinweise
Julabo is a registered trademark of Julabo USA, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)