Z742352
2MAG Stirring bar remover PTFE
flexible with extra strong permanent magnet
Synonym(e):
magnetic extraction tool
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103806
NACRES:
NB.72
Empfohlene Produkte
Hersteller/Markenname
2mag 45070
Verwandte Kategorien
Allgemeine Beschreibung
PTFE-coated flexible stirring bar remover with extremely strong permanent neodym magnet, length 70 cm. Universal chemical resistance also in aggressive media, ideal for fast removal of heavy and big magnetic stirring bars with a weight up to 400 g.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z742352-1EA | 4061824077858 |