Z742345
2MAG mixBATHcover for MIXdrive 6
polycarbonate
Synonym(e):
water bath lid
Alle Fotos(1)
About This Item
UNSPSC-Code:
41103706
NACRES:
NB.72
Empfohlene Produkte
Materialien
polycarbonate
Hersteller/Markenname
2mag 40806
B × T × H
315 mm × 575 mm × 8 mm
Gewicht
1.5 kg
Allgemeine Beschreibung
Chemically resistant, transparent cover for mixBATH, consisting of polycarbonate, for minimization of evaporation effects.
Different cutouts for the perfect positioning of the
thermostat in the case of using the stirring drives
MIXdrive 6 or MIXdrive 15 immersed into the mixBATH.
Different cutouts for the perfect positioning of the
thermostat in the case of using the stirring drives
MIXdrive 6 or MIXdrive 15 immersed into the mixBATH.
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)