Merck
Z740727
Sigma-Aldrich
Aldrich® Essentials boiling flask, flat-bottom, ST/NS 24/29
capacity 50 mL
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass flask
flat bottom
single-neck
Verpackung
pkg of 1 ea
Kapazität
50 mL
Allgemeine Beschreibung
Clear borosilicate glass, flat bottomed boiling flasks with top, outer ST ground joint neck. White marking spot and capacity.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)