Z740240
QianCap™ gas purging set for Ace pressure tubes and bottles
QianCap™-15-v2, fits no. 15 internal thread
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Empfohlene Produkte
Allgemeine Beschreibung
QianCap™ Purging-15v2a can be applied onto bottles and pressure tubes with #15 thread. The set has all items required for immediate use. QianCap purging systems can operate easily, safely, and accurately with a wide array of gases, such as H2, O2, CO2, NH3, N2, Ar, D2, CO, for high reproducibility.
Leistungsmerkmale und Vorteile
- Provided with a blue PTFE face and white silicone backing septum that is disposable.
- Septa is an ideal fit on Q-Tube-12 mL.
- The blue PTFE liner has great chemical resistance.
- Silicon backing has a high resealing ability.
Sonstige Hinweise
Pressure flasks/bottles not included
Rechtliche Hinweise
QianCap is a trademark of Q Labtech LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z740240-1EA | 4061833405451 |