Z689963
Aldrich® column clamp
size L
Alle Fotos(1)
About This Item
UNSPSC-Code:
41122413
NACRES:
NB.35
Empfohlene Produkte
Größe
L
L × Durchm.
211 mm × 11 mm , arm
Backenkapazität
64-140 mm
Verwandte Kategorien
Allgemeine Beschreibung
Corrosion resistant stainless steel. Ideal for holding chromatography columns and large cylindrical glassware. A large, flat thumbscrew/worm drive permits maximum tightening or removal in minimum time.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
NOTE: An additional clamp holder is required to attach clamps to frames or other apparatus.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)