Z689939
Aldrich® flexible support system
with bench clamp, L 457 mm
Alle Fotos(1)
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Empfohlene Produkte
Länge
457 mm
Allgemeine Beschreibung
The unique flexible arms can be placed in virtually any position or angle. Available in three different systems: with steel base plate, with alloy lab frame connector for rods up 19 mm diameter, or aluminum bench clamp.
Each system includes: three nickel-plated clamp heads, 2-prong clamp with 75 mm max. grip, 3-prong clamp with 69 mm max. grip and spring clamp with 13 mm max. grip., and one nickel-plated flex arm in choice of 305 mm length or 457 mm length.
Clamp heads with prongs include vinyl and fiberglass covers for high temperature applications. Utility wrench included with each system.
Each system includes: three nickel-plated clamp heads, 2-prong clamp with 75 mm max. grip, 3-prong clamp with 69 mm max. grip and spring clamp with 13 mm max. grip., and one nickel-plated flex arm in choice of 305 mm length or 457 mm length.
Clamp heads with prongs include vinyl and fiberglass covers for high temperature applications. Utility wrench included with each system.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z689939-1EA | 4061838267757 |