Merck
Z682810
Ace receiving flask,
capacity 500 mL, joint: ST/NS 35/20, glass
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
glass
round bottom flask
single-neck
Verbindung
joint: ST/NS35/20
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Ace Glass 6902-240
Kapazität
500 mL
Allgemeine Beschreibung
The receiving flasks have the standard joint to fit the glassware used in rotary evaporators, and are available with poly-coating or plain glass.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)