Merck
Z682667
Ace drying flask, pear-shaped, with side indents
capacity 1000 mL, joint: ST/NS 24/40, glass
Synonym(e):
Aldrich® drying flask, pear-shaped, with side indents
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
glass
pear-shaped flask
single-neck
Verbindung
joint: ST/NS24/40
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Büchi equivalent, 00424
Kapazität
1000 mL
Allgemeine Beschreibung
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z682667-1EA4061832352602