Merck
Z682527
Ace evaporating flask, pear-shaped
capacity 500 mL, joint: ST/NS 29/42, non-coated, heavy wall
Synonym(e):
Aldrich® evaporating flask, pear-shaped, bottom flask, flask round, round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
pear-shaped flask
single-neck
Verbindung
joint: ST/NS29/42
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Büchi equivalent, 08739
Kapazität
500 mL
Allgemeine Beschreibung
These pear-shaped evaporating flasks can be used as boiling type flasks for rotary evaporators.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)