Merck
Z682330
Ace rotary evaporator condenser complete "A" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Materialien
glass
Hersteller/Markenname
Büchi equivalent, 40633
Verwandte Kategorien
Allgemeine Beschreibung
The complete "A" evaporator condenser assembly includes the main glass condenser body with inner coil for angled installation, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit Büchi® evaporator models 200/205 and series 114-144.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z682330-1EA4061832352053