Merck
Z682179
Ace stopcock only, with inlet feed, for condenser "C" and "CR" assemblies
hollow, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
Verbindung
joint ST/NS19/38
Hersteller/Markenname
Büchi equivalent, 40628
Allgemeine Beschreibung
The Aldrich stopcock fits the Aldrich condenser "C" and "CR" assemblies.
Rechtliche Hinweise
Büchi is a registered trademark of Büchi Labortechnik AG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)