Z680478
Duran® GL 45 connection system caps and accessories
insert for hose connection screw cap GL 14, 3.0 mm
Synonym(e):
1129816
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Empfohlene Produkte
Materialien
white PTFE/silicone
Hersteller/Markenname
DURAN® 1129816
Parameter
-50-230 °C temp. limit
Verwandte Kategorien
Allgemeine Beschreibung
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Sterile pressure equalization is possible through use of membrane filter.
Leistungsmerkmale und Vorteile
- Manufactured from PP, PTFE, silicone
- Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
- Safe and secure liquid transfer into or out of laboratory bottles
- Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
- Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
- Closed-top red PBT GL 14 cap for sealing unused ports
- Sterile pressure equilisation is possible with pressure compensation set
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Kunden haben sich ebenfalls angesehen
Global Trade Item Number
SKU | GTIN |
---|---|
Z680478-1EA | 4061826711460 |