Merck
Z678627
2mag MIX 6 multi position stirrer
AC/DC input 100 / 240 V AC, Schuko plug
Synonym(e):
induction multi-position stirrer, magnetic induction stir plate, magnetic wear-free stirrer, multi-position stirrer
About This Item
UNSPSC-Code:
41103806
NACRES:
NB.72
Materialien
stainless steel plate
Leistungsmerkmale
CE compliant
positions 6
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 30006
Parameter
100-2000 rpm speed
Plattengröße × B × L
× 245 mm × 400 mm
Rührervolumen
1-3000 mL , per stirring position
Allgemeine Beschreibung
Ultra-flat magnetic stirrer with 6 stirring positions, up to 6,000 mL stirring volume, suitable for 6 x 1,000 mL flasks.
Powerful 6 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept, extremely wide speed range of 100 to 2,000 rpm, 100% synchronized speed, jerk-free stirring even at low speeds, 4-step power settings of the stirrer, high power setting for viscous liquids and reduced power setting for long-term operation without any heating effects caused by the stirrer.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Global Trade Item Number
SKUGTIN
Z678627-1EA4061833582596