Merck
Z675423
Replacement mesh inserts for mini-sieve micro sieve set
set of 1
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Verpackung
set of 1
Verwandte Kategorien
Allgemeine Beschreibung
Replacement mesh screens for Mini-Sieve micro sieve set. One each 25, 35, 45, 60, 80, 120, 170 and 230 standard mesh screens.
Die Dokumentenbibliothek aufrufen
Verwandter Inhalt
Particle size conversion table from our Lab Basics Technical Library