Z674370
Duran® GLS 80 caps and accessories
GL 18 pressure equalisation set for 4-port cap
Synonym(e):
1160167
Alle Fotos(1)
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
Empfohlene Produkte
Hersteller/Markenname
DURAN® 1160167
Verwandte Kategorien
Allgemeine Beschreibung
Flexible modular liquid or gas transfer 80 connection cap system. Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be connected. Sterile pressure equalisation is possible by using the syringe membrane filter. Unused ports can be closed with a closed top GL 18 PBT cap.
Leistungsmerkmale und Vorteile
- Manufactured from PP, PTFE, EPDM
- The flexible modular system allows an individual combination of the different components according to requirements and application
- Five different tubing diameters (3.2 mm; 6.0 mm; 8.0 mm; 10.0 mm and 12.0 mm) can be used by changing the inserts
- Sterile pressure equalisation is possible by using the syringe filter
- Unused ports can be sealed off with a blank GL 18 screw cap
- Fully compatible with all the GLS 80 threads and therefore highly versatile
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z674370-1EA | 4061832800981 |