Merck
Z547883
Sigma-Aldrich
SafetyBarb® hose connector
angled
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Allgemeine Beschreibung
Consists of PBT cap, PP barbed connector 8.5 mm O.D., and silicone rubber seal. GL14 thread.
Rechtliche Hinweise
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)