Merck
Z508446
Sigma-Aldrich
Ace flat-bottom reaction flasks
w/o O-ring groove, capacity 1,000 mL
Synonym(e):
bottom flask, flat flask, reaction flasks
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
flat bottom
glass
single-neck
Sterilität
non-sterile
Leistungsmerkmale
closure type Lid
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6511-24
Kapazität
1,000 mL
Allgemeine Beschreibung
Cylindrical heavy-wall reaction flask with a flat bottom rounded into side of wall, and extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Optional O-ring groove allows use of CAPFE O-rings rather than gaskets. Flask is supplied with one CAPFE O-ring. Order reaction flask heads and clamps separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)